Assay Details
| Gene Symbol: LOC100067608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014737740.2 | XP_014593226.2 | 2888 | 85 | ||
| XM_023636336.1 | XP_023492104.1 | 2888 | 85 | |||
More Information
Set Membership: |
|

LOC100067608
Genomic Map