Assay Details
| Gene Symbol: C2H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607794.3 | XP_005607851.1 | 1169 | 88 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C2H4orf45| Gene Symbol: C2H4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005607794.3 | XP_005607851.1 | 1169 | 88 | ||