Assay Details
| Gene Symbol: RPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502992.4 | XP_001503042.2 | 394 | 93 | ||
| XM_003364529.4 | XP_003364577.1 | 356 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RPL34| Gene Symbol: RPL34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502992.4 | XP_001503042.2 | 394 | 93 | ||
| XM_003364529.4 | XP_003364577.1 | 356 | 93 | |||
Set Membership: |
|