Assay Details
| Gene Symbol: SERPINB9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734698.2 | XP_014590184.1 | 158 | 117 | ||
| XM_023624361.1 | XP_023480129.1 | 158 | 117 | |||
| XM_023624362.1 | XP_023480130.1 | 158 | 117 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014734698.2, XM_023624361.1, XM_023624362.1 transcripts.
Set Membership: |
|

SERPINB9
Genomic Map