Assay Details
| Gene Symbol: RIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003363744.4 | XP_003363792.2 | 580 | 106 | ||
| XM_005603606.3 | XP_005603663.2 | 577 | 106 | |||
| XM_014734222.2 | XP_014589708.1 | 474 | 106 | |||
| XM_023624535.1 | XP_023480303.1 | 520 | 106 | |||
| XM_023624536.1 | XP_023480304.1 | 385 | 106 | |||
More Information
Set Membership: |
|

RIPOR2
Genomic Map