Assay Details
| Gene Symbol: LOC100067842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624544.1 | XP_023480312.1 | 1818 | 125 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100067842| Gene Symbol: LOC100067842 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023624544.1 | XP_023480312.1 | 1818 | 125 | ||
Set Membership: |
|