Assay Details
| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603633.3 | XP_005603690.1 | 296 | 102 | ||
| XM_014734262.2 | XP_014589748.1 | 360 | 102 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005603633.3, XM_014734262.2 transcripts.
Set Membership: |
|

ZNF184
Genomic Map