Assay Details
| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603633.3 | XP_005603690.1 | 489 | 85 | ||
| XM_014734262.2 | XP_014589748.1 | 553 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF184| Gene Symbol: ZNF184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603633.3 | XP_005603690.1 | 489 | 85 | ||
| XM_014734262.2 | XP_014589748.1 | 553 | 85 | |||
Set Membership: |
|