Assay Details
| Gene Symbol: POLR1H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603662.3 | XP_005603719.1 | 151 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005603662.3 transcripts.
Set Membership: |
|

POLR1H
Genomic Map