Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917925.5 | XP_001917960.3 | 1788 | 129 | ||
| XM_023624835.1 | XP_023480603.1 | 969 | 129 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001917925.5 | XP_001917960.3 | 1788 | 129 | ||
| XM_023624835.1 | XP_023480603.1 | 969 | 129 | |||
Set Membership: |
|