Assay Details
| Gene Symbol: LOC100066125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603881.3 | XP_005603938.1 | 781 | 110 | ||
| XM_005603882.3 | XP_005603939.1 | 781 | 110 | |||
| XM_005603883.3 | XP_005603940.1 | 780 | 110 | |||
| XM_014734399.2 | XP_014589885.1 | 781 | 110 | |||
| XM_014734400.2 | XP_014589886.1 | 590 | 110 | |||
| XM_023624889.1 | XP_023480657.1 | 785 | 110 | |||
| XM_023624890.1 | XP_023480658.1 | 785 | 110 | |||
| XM_023624891.1 | XP_023480659.1 | 785 | 110 | |||
More Information
Set Membership: |
|

LOC100066125
Genomic Map