Assay Details
| Gene Symbol: PRICKLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005603908.3 | XP_005603965.1 | 561 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005603908.3 transcripts.
Set Membership: |
|

PRICKLE4
Genomic Map