Assay Details
| Gene Symbol: BICRAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734429.2 | XP_014589915.1 | 225 | 94 | ||
| XM_023624943.1 | XP_023480711.1 | 810 | 94 | |||
| XM_023624944.1 | XP_023480712.1 | 577 | 94 | |||

BICRAL
Genomic Map