Assay Details
| Gene Symbol: CILK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918274.4 | XP_001918309.1 | 2192 | 72 | ||
| XM_023625092.1 | XP_023480860.1 | 2287 | 72 | |||
| XM_023625094.1 | XP_023480862.1 | 1693 | 72 | |||
More Information
Set Membership: |
|

CILK1
Genomic Map