Assay Details
| Gene Symbol: CILK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001918274.4 | XP_001918309.1 | 275 | 78 | ||
| XM_023625093.1 | XP_023480861.1 | 276 | 78 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001918274.4, XM_023625093.1 transcripts.
Set Membership: |
|

CILK1
Genomic Map