Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503391.4 | XP_001503441.1 | 804 | 104 | ||
| XM_014734785.2 | XP_014590271.1 | 882 | 104 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CERS1| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001503391.4 | XP_001503441.1 | 804 | 104 | ||
| XM_014734785.2 | XP_014590271.1 | 882 | 104 | |||
Set Membership: |
|