Assay Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604230.3 | XP_005604287.1 | 187 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005604230.3 transcripts.
Set Membership: |
|

SREK1IP1
Genomic Map