Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604289.3 | XP_005604346.1 | 271 | 84 | ||
| XM_023625685.1 | XP_023481453.1 | 274 | 84 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL28| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604289.3 | XP_005604346.1 | 271 | 84 | ||
| XM_023625685.1 | XP_023481453.1 | 274 | 84 | |||
Set Membership: |
|