Assay Details
| Gene Symbol: ZNF131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625689.1 | XP_023481457.1 | 456 | 136 | ||
| XM_023625690.1 | XP_023481458.1 | 460 | 136 | |||
| XM_023625691.1 | XP_023481459.1 | 451 | 136 | |||
| XM_023625692.1 | XP_023481460.1 | 580 | 136 | |||
| XM_023625693.1 | XP_023481461.1 | 540 | 136 | |||
| XM_023625694.1 | XP_023481462.1 | 455 | 136 | |||
More Information
Set Membership: |
|

ZNF131
Genomic Map