Assay Details
| Gene Symbol: RPL37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498955.6 | XP_001499005.1 | 148 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001498955.6 transcripts.
Set Membership: |
|

RPL37
Genomic Map