Assay Details
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604343.3 | XP_005604400.1 | 177 | 96 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005604343.3 transcripts.
Set Membership: |
|

DAP
Genomic Map