Assay Details
| Gene Symbol: MARCHF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014734935.2 | XP_014590421.2 | 937 | 85 | ||
| XM_023625849.1 | XP_023481617.1 | 937 | 85 | |||
| XM_023625850.1 | XP_023481618.1 | 1086 | 85 | |||
More Information
Set Membership: |
|

MARCHF6
Genomic Map