Assay Details
| Gene Symbol: ATPSCKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625851.1 | XP_023481619.1 | 325 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATPSCKMT| Gene Symbol: ATPSCKMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023625851.1 | XP_023481619.1 | 325 | 91 | ||
Set Membership: |
|