Assay Details
| Gene Symbol: TENT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604354.3 | XP_005604411.1 | 865 | 85 | ||
| XM_005604355.3 | XP_005604412.1 | 867 | 85 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TENT4A| Gene Symbol: TENT4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604354.3 | XP_005604411.1 | 865 | 85 | ||
| XM_005604355.3 | XP_005604412.1 | 867 | 85 | |||
Set Membership: |
|