Assay Details
| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643246.1 | XP_023499014.1 | 554 | 86 | ||
| GenBank mRNA | JL640328.1 | - | 5 - 6 | 515 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IRAG2| Gene Symbol: IRAG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023643246.1 | XP_023499014.1 | 554 | 86 | ||
| GenBank mRNA | JL640328.1 | - | 5 - 6 | 515 | 86 | |
Set Membership: |
|