Assay Details
| Gene Symbol: LOC100070179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611059.3 | XP_005611116.2 | 1576 | 82 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100070179| Gene Symbol: LOC100070179 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005611059.3 | XP_005611116.2 | 1576 | 82 | ||
Set Membership: |
|