Assay Details
| Gene Symbol: LOC100051901 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504181.4 | XP_001504231.1 | 130 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001504181.4 transcripts.
Set Membership: |
|

LOC100051901
Genomic Map