Assay Details
| Gene Symbol: RBBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490566.4 | XP_001490616.1 | 330 | 80 | ||
| XM_023626049.1 | XP_023481817.1 | 483 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBBP9| Gene Symbol: RBBP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001490566.4 | XP_001490616.1 | 330 | 80 | ||
| XM_023626049.1 | XP_023481817.1 | 483 | 80 | |||