Assay Details
| Gene Symbol: LZTS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626207.1 | XP_023481975.1 | 861 | 100 | ||
| XM_023626208.1 | XP_023481976.1 | 772 | 100 | |||
| XM_023626209.1 | XP_023481977.1 | 1039 | 100 | |||
| XM_023626210.1 | XP_023481978.1 | 861 | 100 | |||
| XM_023626211.1 | XP_023481979.1 | 923 | 100 | |||
| XM_023626212.1 | XP_023481980.1 | 882 | 100 | |||
| XM_023626213.1 | XP_023481981.1 | 885 | 100 | |||
More Information
Set Membership: |
|

LZTS3
Genomic Map