Assay Details
| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500940.4 | XP_001500990.1 | 275 | 76 | ||
| XM_005604574.3 | XP_005604631.1 | 276 | 76 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PXMP4| Gene Symbol: PXMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500940.4 | XP_001500990.1 | 275 | 76 | ||
| XM_005604574.3 | XP_005604631.1 | 276 | 76 | |||