Assay Details
| Gene Symbol: C22H20orf173 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604599.2 | XP_005604656.1 | 633 | 71 | ||
| XM_023625938.1 | XP_023481706.1 | 633 | 71 | |||
More Information
Set Membership: |
|

C22H20orf173
Genomic Map