Assay Details
| Gene Symbol: LOC100070683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626596.1 | XP_023482364.1 | 4895 | 91 | ||
| XM_023626597.1 | XP_023482365.1 | 1783 | 91 | |||
| XM_023626599.1 | XP_023482367.1 | 4895 | 91 | |||
| XM_023626600.1 | XP_023482368.1 | 254 | 91 | |||
More Information
Set Membership: |
|

LOC100070683
Genomic Map