Assay Details
| Gene Symbol: LOC100070893 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604653.3 | XP_005604710.1 | 309 | 67 | ||
| XM_005604654.3 | XP_005604711.1 | 309 | 67 | |||
More Information
Set Membership: |
|

LOC100070893
Genomic Map