Assay Details
| Gene Symbol: TP53TG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500634.4 | XP_001500684.1 | 372 | 76 | ||
| XM_023626624.1 | XP_023482392.1 | 496 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TP53TG5| Gene Symbol: TP53TG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001500634.4 | XP_001500684.1 | 372 | 76 | ||
| XM_023626624.1 | XP_023482392.1 | 496 | 76 | |||
Set Membership: |
|