Assay Details
| Gene Symbol: TMEM189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604709.3 | XP_005604766.2 | 248 | 51 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM189| Gene Symbol: TMEM189 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005604709.3 | XP_005604766.2 | 248 | 51 | ||
Set Membership: |
|