Assay Details
| Gene Symbol: LOC100061341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495873.5 | XP_001495923.1 | 2242 | 81 | ||
| XM_005604939.3 | XP_005604996.1 | 2272 | 81 | |||
| XM_014735401.2 | XP_014590887.1 | 2020 | 81 | |||
More Information
Set Membership: |
|

LOC100061341
Genomic Map