Assay Details
| Gene Symbol: LOC100061341 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495873.5 | XP_001495923.1 | 140 | 87 | ||
| XM_005604939.3 | XP_005604996.1 | 140 | 87 | |||
| XM_014735401.2 | XP_014590887.1 | 140 | 87 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001495873.5, XM_005604939.3, XM_014735401.2 transcripts.
Set Membership: |
|

LOC100061341
Genomic Map