Assay Details
| Gene Symbol: LOC100058496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627281.1 | XP_023483049.1 | 426 | 106 | ||
| XM_023627282.1 | XP_023483050.1 | 427 | 106 | |||
| XM_023627283.1 | XP_023483051.1 | 427 | 106 | |||
More Information
Set Membership: |
|

LOC100058496
Genomic Map