Assay Details
| Gene Symbol: LOC100058496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023627281.1 | XP_023483049.1 | 535 | 71 | ||
| XM_023627282.1 | XP_023483050.1 | 536 | 71 | |||
| XM_023627283.1 | XP_023483051.1 | 536 | 71 | |||
More Information
Set Membership: |
|

LOC100058496
Genomic Map