Assay Details
| Gene Symbol: LOC100064891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001495656.5 | XP_001495706.2 | 122 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001495656.5 transcripts.
Set Membership: |
|

LOC100064891
Genomic Map