Assay Details
| Gene Symbol: TEDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628575.1 | XP_023484343.1 | 670 | 90 | ||
| XM_023628576.1 | XP_023484344.1 | 487 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEDC1| Gene Symbol: TEDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023628575.1 | XP_023484343.1 | 670 | 90 | ||
| XM_023628576.1 | XP_023484344.1 | 487 | 90 | |||