Assay Details
| Gene Symbol: ABITRAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492457.4 | XP_001492507.1 | 202 | 72 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABITRAM| Gene Symbol: ABITRAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001492457.4 | XP_001492507.1 | 202 | 72 | ||