Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605708.3 | XP_005605765.2 | 638 | 92 | ||
| XM_014735970.2 | XP_014591456.1 | 384 | 92 | |||
| XM_023628672.1 | XP_023484440.1 | 1153 | 92 | |||
| XM_023628673.1 | XP_023484441.1 | 385 | 92 | |||
More Information
Set Membership: |
|

ZFP37
Genomic Map