Assay Details
| Gene Symbol: LOC102149958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014736009.2 | XP_014591495.1 | 2361 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014736009.2 transcripts.
Set Membership: |
|

LOC102149958
Genomic Map