Assay Details
| Gene Symbol: LOC102149958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005605748.3 | XP_005605805.1 | 662 | 92 | ||
| XM_014736009.2 | XP_014591495.1 | 2783 | 92 | |||
| XM_014736010.2 | XP_014591496.1 | 2670 | 92 | |||
| XM_023628771.1 | XP_023484539.1 | 822 | 92 | |||
More Information
Set Membership: |
|

LOC102149958
Genomic Map