Assay Details
| Gene Symbol: RPL35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001502048.6 | XP_001502098.3 | 153 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001502048.6 transcripts.
Set Membership: |
|

RPL35
Genomic Map