Assay Details
| Gene Symbol: BLOC1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504775.5 | XP_001504825.1 | 377 | 74 | ||
| XM_023643751.1 | XP_023499519.1 | 332 | 74 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BLOC1S1| Gene Symbol: BLOC1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001504775.5 | XP_001504825.1 | 377 | 74 | ||
| XM_023643751.1 | XP_023499519.1 | 332 | 74 | |||
Set Membership: |
|