Assay Details
| Gene Symbol: LOC100069029 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498857.5 | XP_001498907.4 | 371 | 59 | ||
| XM_023629287.1 | XP_023485055.1 | 371 | 59 | |||
More Information
Set Membership: |
|

LOC100069029
Genomic Map