Assay Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498210.6 | XP_001498260.5 | 196 | 73 | ||
| XM_023629375.1 | XP_023485143.1 | 196 | 73 | |||
| XM_023629376.1 | XP_023485144.1 | 584 | 73 | |||
| XM_023629377.1 | XP_023485145.1 | 574 | 73 | |||
More Information
Set Membership: |
|

GLT6D1
Genomic Map