Assay Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498210.6 | XP_001498260.5 | 245 | 62 | ||
| XM_023629377.1 | XP_023485145.1 | 623 | 62 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GLT6D1| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001498210.6 | XP_001498260.5 | 245 | 62 | ||
| XM_023629377.1 | XP_023485145.1 | 623 | 62 | |||
Set Membership: |
|